Learn More
Carbosynth LLC. Tachyplesin III, 1mg, MF= C99H151N33O19S4
SDP
Supplier:  Carbosynth LLC. TAC3046PI1MG
Encompass_Preferred
H-Lys-Trp-Cys-Phe-Arg-Val-Cys-Tyr-Arg-Gly-Ile-Cys-Tyr-Arg-Lys-Cys-Arg-NH2, Application: Antimicrobial Peptide, Notes: Tachyplesin is a type of cationic ß-hairpin antimicrobial peptide (AMP) discovered from horseshoe crab hemocytes., salt: Trifluoroacetate Salt, one letter code: H-KWCFRVCYRGICYRKCR-NH2, MW: 2235.77, Disulfide bonds: Disulfide Bonds Between Cys3-Cys16,Cys7-Cys12
Catalog No. 50-195-2437