Learn More
AdipoGen TACI (human):Fc (human) (rec.) (50 µg)
SDP
Supplier:  AdipoGen AG40B0079C050
Encompass_Preferred
Protein. The extracellular domain of human TACI (aa 2-118) is fused at the C-terminus to the Fc portion of human IgG1. Source: HEK 293 cells. Endotoxin content: <0.01EU/µg purified protein (LAL test; Lonza). Lyophilized. Contains PBS. Binds to human and mouse BAFF and APRIL. Purity: >90% (SDS-PAGE). B-cell activating factor belonging to the TNF family receptor (BAFF-R), B-cell maturation antigen (BCMA), and transmembrane activator and calcium modulator and cyclophilin ligand interactor (TACI) play critical functions in promoting B-cell survival by engaging the ligands a proliferation-inducing ligand (APRIL) and/or BAFF. TNFRSF13B, the gene, which encodes TACI is mutated in nearly 10% of patients with common variable immune deficiency (CVID), an antibody deficiency syndrome characterized by loss of memory B cells and plasma cells.
Catalog No. 50-205-3141