Learn More
Enzo Life Sciences TACI (mouse) monoclonal antibody (8F10) (100µg)
SDP
Supplier:  Enzo Life Sciences ALX804800C100
Encompass_Preferred
Clone: 8F10. Isotype: IgG2a. Immunogen: Rat RBL cells expressing mouse TACI. Source: Purified from concentrated hybridoma tissue culture supernatant. Species reactivity: Mouse. Applications: Flow Cytometry, IHC (FS), FUNC. Formulation: Liquid. In PBS.
Catalog No. 50-200-9970