Learn More
AdipoGen anti-TACI (mouse), mAb (1A-10) (Biotin) (100 µg)
SDP
Supplier:  AdipoGen AG20B0035BC100
Encompass_Preferred
Monoclonal Antibody. Recognizes mouse TACI. Isotype: Rat IgG2a. Clone: 1A-10. Applications: FACS. Liquid. In PBS containing 0.02% sodium azide. Three receptors for BAFF were recently identified and designated TACI, BCMA and BAFF-R. BAFF-R signaling is essential for B cell development, survival and humoral immune responses.
Catalog No. 50-205-2939