Learn More
Enzo Life Sciences TACI polyclonal antibody (100µg)
SDP
Supplier:  Enzo Life Sciences ADI905312100
Encompass_Preferred
Host: Rabbit. Immunogen: Synthetic peptide corresponding to the sequence near the N-terminus of human TACI. UniProt ID: O14836. Species reactivity: Human. Applications: ICC, WB. Recommended Dilutions: Immunocytochemistry (2µg/ml) Western Blot (2-5µg/ml) Suggested dilutions/conditions may not be available for all applications. Optimal conditions must be determined individually for each application. Purity: Peptide affinity purified. Formulation: Liquid. In PBS containing 0.02% sodium azide.
Catalog No. 50-200-6966