Learn More
ACROBiosystems Recombinant Protein;Biotinylated Human TACI / TNFRSF13B Protein, Fc Tag, Avi Tag (Avitag™);HEK293;200UG;TAI,CD267,TACI,TNFRSF13B
Catalog No. 501657859
Encompass_Preferred
Recombinant Protein;Biotinylated Human TACI / TNFRSF13B Protein, Fc Tag, Avi Tag (Avitag™);200UG;Biotinylated Human TACI, Fc Tag, Avi Tag (TAI-H82F6) is expressed from human 293 cells (HEK293). It contains AA Ser 2 - Thr 166 (Accession # O14836-1).;This protein carries a human IgG1 Fc tag at the C-terminus, followed by a Avi tag (Avitag™). The protein has a calculated MW of 46.8 kDa. The protein migrates as 33-40 kDa and 47-50 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.;Immobilized Human BAFF, Fc Tag (Cat. No. BAF-H4268) at 2 μg/mL (100 μL/well) can bind Biotinylated Human TACI, Fc Tag, Avi Tag (Cat. No. TAI-H82F6) with a linear range of 1.2-39 ng/ml (QC tested).
Catalog No. 50-165-7859 Supplier ACROBiosystems Supplier No. TAIH82F6200UG
Recombinant Protein;Biotinylated Human TACI / TNFRSF13B Protein, Fc Tag, Avi Tag (Avitag™);200UG;Biotinylated Human TACI, Fc Tag, Avi Tag (TAI-H82F6) is expressed from human 293 cells (HEK293). It contains AA Ser 2 - Thr 166 (Accession # O14836-1).;This protein carries a human IgG1 Fc tag at the C-terminus, followed by a Avi tag (Avitag™). The protein has a calculated MW of 46.8 kDa. The protein migrates as 33-40 kDa and 47-50 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.;Immobilized Human BAFF, Fc Tag (Cat. No. BAF-H4268) at 2 μg/mL (100 μL/well) can bind Biotinylated Human TACI, Fc Tag, Avi Tag (Cat. No. TAI-H82F6) with a linear range of 1.2-39 ng/ml (QC tested).