Learn More
MSC Gerson Tack Cloth, Synthetic, 9 x 18 in, Blue, 12/Box
Catalog No. 502869141
Encompass
Blend Prep™ Tack Cloths are engineered for optimal surface preparation in painting applications. These cloths utilize synthetic hydro-entangled fibers, ensuring a durable, tear-resistant, and lint-free fabric. Their apertured design efficiently picks up and traps particulates, removing surface contaminants completely without leaving harmful residues.
  • Dry tack
  • Synthetic hydro-entangled fibers provide durable, tear-resistant, and lint-free fabric
  • Apertured design effectively picks up and traps all particulates
  • Completely removes surface particulates without leaving harmful residues
  • Designed for waterborne paints; compatible with all paint types
Catalog No. 50-286-9141 Supplier MSC Supplier No. 86456365
Blend Prep™ Tack Cloths are engineered for optimal surface preparation in painting applications. These cloths utilize synthetic hydro-entangled fibers, ensuring a durable, tear-resistant, and lint-free fabric. Their apertured design efficiently picks up and traps particulates, removing surface contaminants completely without leaving harmful residues.
  • Dry tack
  • Synthetic hydro-entangled fibers provide durable, tear-resistant, and lint-free fabric
  • Apertured design effectively picks up and traps all particulates
  • Completely removes surface particulates without leaving harmful residues
  • Designed for waterborne paints; compatible with all paint types