Learn More
Medchemexpress LLC HY-B2244 100mg Medchemexpress, Tacrine hydrochloride hydrate CAS:206658-92-6 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HYB2244100MG
Encompass_Preferred
Medchemexpress, HY-B2244 100mg Tacrine hydrochloride hydrate CAS:206658-92-6 Tacrine hydrochloride hydrate is an inhibitor of both acetyl ( AChE ) and butyryl-cholinestrase ( BChE ) with IC 50 s of 31 nM and 25.6 nM, respectively. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-193-3127