Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tacrine hydrochloride

Catalog No. aaj6007006
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAJ6007006 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAJ6007006 Supplier Thermo Scientific Chemicals Supplier No. J6007006
Only null left
Add to Cart

A potent centrally acting anticholinesterase | CAS: 1684-40-8 | C13H15ClN2 | 234.73 g/mol

A potent centrally acting anticholinesterase for therapy of memory deficits in patients with Alzheimer's disease. THA also selectively blocks potassium channels in the central nervous system, which results in an increased release of acetylcholine and a prolongation of the action potential of the presynaptic cholinergic neurons.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Solubility
Soluble in aqueous buffers

Notes
Refrigerate. Keep container tightly closed in a dry and well-ventilated place. Store away from oxidizing agent.
TRUSTED_SUSTAINABILITY

Specifications

CAS 1684-40-8
Melting Point 283°C to 284°C
Color Yellow
Molecular Formula C13H15ClN2
Quantity 5 g
MDL Number MFCD00150067
UN Number UN2811
Merck Index 14,9024
Synonym tacrine hydrochloride, 1,2,3,4-tetrahydroacridin-9-amine hydrochloride, tacrine hcl, hydroaminacrine, tenakrin, 9-amino-1,2,3,4-tetrahydroacridine hydrochloride, tacrine.hcl, unii-4966rng0bu, 1,2,3,4-tetrahydro-9-acridinamine hydrochloride, ccris 7250
Solubility Information Soluble in aqueous buffers
InChI Key ZUFVXZVXEJHHBN-UHFFFAOYSA-N
SMILES [H+].[Cl-].NC1=C2C=CC=CC2=NC2=C1CCCC2
IUPAC Name hydrogen 1,2,3,4-tetrahydroacridin-9-amine chloride
Molecular Weight (g/mol) 234.73
PubChem CID 2723754
Formula Weight 234.73 (Anhydrous)
Percent Purity ≥98%
Physical Form Crystalline Solid
Chemical Name or Material Tacrine hydrochloride
Show More Show Less
Hazard Category H302+H332-H315-H319-H335
Hazard Statement GHS H Statement
H331-H302-H315-H319-H335
Toxic if inhaled.
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
DOTInformation Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC SOLIDS, ORGANIC, N.O.S.
EINECSNumber 216-867-5
RTECSNumber AR9532500
TSCA No
Recommended Storage Keep cold

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.