Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Tacrine hydrochloride | 1684-40-8 | 99.9% | 1 ML
SDP

Catalog No. 5000411131
Encompass_Preferred

Tacrine hydrochloride is a potent brain-penetrant inhibitor of both AChE and BChE. It also acts as an NMDAR inhibitor. This compound can be utilized in research related to Alzheimer's disease.

  • Potent brain-penetrant inhibitor of AChE and BChE.
  • NMDAR inhibitor.
  • Used for Alzheimer's disease research.
  • Appearance: Solid, white to light yellow.
  • Shipping: Room temperature in continental US.
  • Storage: 4°C, stored under nitrogen, away from moisture.
  • In solvent storage: -80°C for 6 months; -20°C for 1 month (stored under nitrogen, away from moisture).
  • Solubility in H2O: 33.33 mg/mL (141.99 mM; needs ultrasonic).

Catalog No. 50-004-11131 Supplier Medchemexpress LLC Supplier No. HYB148810MM/1ML
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Tacrine hydrochloride is a potent brain-penetrant inhibitor of both AChE and BChE. It also acts as an NMDAR inhibitor. This compound can be utilized in research related to Alzheimer's disease.

  • Potent brain-penetrant inhibitor of AChE and BChE.
  • NMDAR inhibitor.
  • Used for Alzheimer's disease research.
  • Appearance: Solid, white to light yellow.
  • Shipping: Room temperature in continental US.
  • Storage: 4°C, stored under nitrogen, away from moisture.
  • In solvent storage: -80°C for 6 months; -20°C for 1 month (stored under nitrogen, away from moisture).
  • Solubility in H2O: 33.33 mg/mL (141.99 mM; needs ultrasonic).

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.