Learn More
Medchemexpress LLC Tacrolimus | 104987-11-3 | MFCD00869680 | ≥98% | 804.01 | 1 G
SDP
Catalog No. 5000352274
Encompass_Preferred
Tacrolimus (FK506) is a macrocyclic lactone that binds to FK506 binding protein (FKBP) to form a complex. It inhibits calcineurin phosphatase, which in turn inhibits T-lymphocyte signal transduction and IL-2 transcription, exhibiting immunosuppressive properties.
  • Binds to FK506 binding protein (FKBP)
  • Inhibits calcineurin phosphatase
  • Inhibits T-lymphocyte signal transduction
  • Inhibits IL-2 transcription
  • Possesses immunosuppressive properties
Catalog No. 50-003-52274 Supplier Medchemexpress LLC Supplier No. HY137561G
Tacrolimus (FK506) is a macrocyclic lactone that binds to FK506 binding protein (FKBP) to form a complex. It inhibits calcineurin phosphatase, which in turn inhibits T-lymphocyte signal transduction and IL-2 transcription, exhibiting immunosuppressive properties.
  • Binds to FK506 binding protein (FKBP)
  • Inhibits calcineurin phosphatase
  • Inhibits T-lymphocyte signal transduction
  • Inhibits IL-2 transcription
  • Possesses immunosuppressive properties