Learn More
Medchemexpress LLC Tacrolimus | 104987-11-3 | MFCD00869853 | >98.0% | 804.02 | C44H69NO12 | 5 MG
SDP
Catalog No. 5000221964
Encompass_Preferred
Tacrolimus is a macrolide immunosuppressant (FK506) used in research to inhibit calcineurin-dependent T-cell activation. It binds FK506-binding protein to form a complex that inhibits calcineurin phosphatase, reducing IL-2 transcription and downstream immune responses. Supplied as a high-purity analytical standard for research and analytical applications.
  • Inhibits calcineurin-dependent T-cell activation.
  • Binds FK506-binding proteins to form an inhibitory complex.
  • High purity (>98%) suitable for analytical work.
  • Molecular weight 804.02, formula C44H69NO12.
  • Useful for immunology and signal transduction studies.
  • Provided in small research quantities appropriate for assays.
Catalog No. 50-002-21964 Supplier Medchemexpress LLC Supplier No. HY137565MG
Tacrolimus is a macrolide immunosuppressant (FK506) used in research to inhibit calcineurin-dependent T-cell activation. It binds FK506-binding protein to form a complex that inhibits calcineurin phosphatase, reducing IL-2 transcription and downstream immune responses. Supplied as a high-purity analytical standard for research and analytical applications.
  • Inhibits calcineurin-dependent T-cell activation.
  • Binds FK506-binding proteins to form an inhibitory complex.
  • High purity (>98%) suitable for analytical work.
  • Molecular weight 804.02, formula C44H69NO12.
  • Useful for immunology and signal transduction studies.
  • Provided in small research quantities appropriate for assays.