Learn More
Medchemexpress LLC Tacrolimus | 104987-11-3 | MFCD00869853 | 99.6% | C44H69NO12 | 10MG
SDP
Catalog No. 5000259432
Encompass_Preferred
Tacrolimus (Standard) is an analytical reference standard of tacrolimus (FK506) intended for research and analytical laboratories. It is used for calibration, method development, and qualitative or quantitative analyses by techniques such as HPLC, GC, and mass spectrometry, and is supplied with supporting documentation for analytical use.
  • High purity (99.6%) suitable for trace-level quantitation.
  • Compatible with HPLC, GC, and MS analytical methods.
  • Supplied as an analytical standard for calibration and method development.
  • Certificate of analysis and safety data sheet available.
  • Available in multiple small pack sizes for analytical workflows.
Catalog No. 50-002-59432 Supplier Medchemexpress LLC Supplier No. HY13756R10MG
Tacrolimus (Standard) is an analytical reference standard of tacrolimus (FK506) intended for research and analytical laboratories. It is used for calibration, method development, and qualitative or quantitative analyses by techniques such as HPLC, GC, and mass spectrometry, and is supplied with supporting documentation for analytical use.
  • High purity (99.6%) suitable for trace-level quantitation.
  • Compatible with HPLC, GC, and MS analytical methods.
  • Supplied as an analytical standard for calibration and method development.
  • Certificate of analysis and safety data sheet available.
  • Available in multiple small pack sizes for analytical workflows.