Learn More
Medchemexpress LLC HY-13756 500mg , Tacrolimus Fujimycin;FR900506;FK506 CAS:104987-11-3 Purity:98%
SDP
Supplier:  Medchemexpress LLC HY13756500MG
Encompass_Preferred
Medchemexpress, HY-13756 500mg Tacrolimus Fujimycin;FR900506;FK506 CAS:104987-11-3 Formula:C44H69NO12 PP2B (calcineurin phosphatase) Autophagy inducer Purity:98% Tacrolimus binds to FK506 binding protein ( FKBP ). This complex inhibits calcineurin phosphatase (PP2B). Tacrolimus is a mTOR-independent autophagy inducer. Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-149-2324