Learn More
TARGETMOL CHEMICALS INC Tacrolimus 50mg
SDP
Catalog No. NC1678288
Encompass_Preferred
Tacrolimus can bind FKBP12 to form a high-affinity complex (Ki: 0.2 nM) which inhibits the activity of the calcium/calmodulin-dependent protein phosphatase.
Catalog No. NC1678288 Supplier TARGETMOL CHEMICALS INC Supplier No. T214450MG
Tacrolimus can bind FKBP12 to form a high-affinity complex (Ki: 0.2 nM) which inhibits the activity of the calcium/calmodulin-dependent protein phosphatase.