Learn More
MOLECULAR DEPOT LLC Tacrolimus Beads - 1 mL
Catalog No. 5000013481
Encompass_Preferred
Tacrolimus Antibody Conjugated to Magnetic Beads consists of magnetic particles (1 um in diameter 0 1perc solid) conjugated to Tacrolimus though a (PEG)n linker This product has been used as tools for the research and development of CLIA methods for the detection of Tacrolimus This product is for in vitro research use only
Catalog No. 50-000-13481 Supplier MOLECULAR DEPOT LLC Supplier No. B2010391
Tacrolimus Antibody Conjugated to Magnetic Beads consists of magnetic particles (1 um in diameter 0 1perc solid) conjugated to Tacrolimus though a (PEG)n linker This product has been used as tools for the research and development of CLIA methods for the detection of Tacrolimus This product is for in vitro research use only