Learn More
Medchemexpress LLC Tacrolimus/BSA | 1 MG
SDP
Catalog No. 5000393128
Encompass_Preferred
Tacrolimus/BSA is an antigen-adjuvant conjugate formed by conjugating Tacrolimus with Bovine Serum Albumin (BSA). This conjugation enhances the production of antigen-specific antibodies in vaccine models. It does not affect protein folding or disrupt major epitopes, and it can enhance cross-presentation and the generation of antigen-specific T cells.
  • Enhances production of antigen-specific antibodies
  • Does not affect protein folding
  • Does not disrupt major epitopes
  • Enhances cross-presentation
  • Generates antigen-specific T cells
Catalog No. 50-003-93128 Supplier Medchemexpress LLC Supplier No. HY1615601MG
Tacrolimus/BSA is an antigen-adjuvant conjugate formed by conjugating Tacrolimus with Bovine Serum Albumin (BSA). This conjugation enhances the production of antigen-specific antibodies in vaccine models. It does not affect protein folding or disrupt major epitopes, and it can enhance cross-presentation and the generation of antigen-specific T cells.
  • Enhances production of antigen-specific antibodies
  • Does not affect protein folding
  • Does not disrupt major epitopes
  • Enhances cross-presentation
  • Generates antigen-specific T cells