Learn More
MOLECULAR DEPOT LLC Tacrolimus BSA - 1 mg
Catalog No. 5000013493
Encompass_Preferred
Tacrolimus BSA Conjugate Solution is a concentrated solution of Bovine Serum Albumin (BSA) conjugated to Tacrolimus (FK506) A carbon spacer arm links BSA to the Tacrolimus moiety Each BSA molecule contains several Tacrolimus units Dilutions of the provided conjugate can be made with Phosphate Buffer Saline For in vitro research use only
Catalog No. 50-000-13493 Supplier MOLECULAR DEPOT LLC Supplier No. P2010001
Tacrolimus BSA Conjugate Solution is a concentrated solution of Bovine Serum Albumin (BSA) conjugated to Tacrolimus (FK506) A carbon spacer arm links BSA to the Tacrolimus moiety Each BSA molecule contains several Tacrolimus units Dilutions of the provided conjugate can be made with Phosphate Buffer Saline For in vitro research use only