Learn More
MOLECULAR DEPOT LLC Tacrolimus Buffer - 25 mL
Catalog No. 5000013425
Encompass_Preferred
Tacrolimus Release Buffer is an extraction solution for the release of tacrolimus from its protein-bound form This product has been used as a tool for the research of detection systems for Tacrolimus Store at 2-8C For in vitro research use only
Catalog No. 50-000-13425 Supplier MOLECULAR DEPOT LLC Supplier No. B2010730
Tacrolimus Release Buffer is an extraction solution for the release of tacrolimus from its protein-bound form This product has been used as a tool for the research of detection systems for Tacrolimus Store at 2-8C For in vitro research use only