Learn More
Apexbio Technology LLC Tacrolimus (FK506), 100mg. Cas: 104987-11-3 MFCD: MFCD00869853. Macrolide calcineurin inhibitor,immunosuppressant
SDP
Supplier:  Apexbio Technology LLC B2143100
Encompass_Preferred
Tacrolimus (FK506) is a potent and selective inhibitor of T-lymphocyte and the macrolide immunosuppressant [1].In vitro, Tacrolimus (FK506) has been reported to inhibit the interleukin 2(IL-2) production and the response of mixed lymphocyte culture. Other sizes are available. Please inqury us for quote.
Catalog No. 50-190-5317