Learn More
Apexbio Technology LLC Tacrolimus (FK506), 104987-11-3, MFCD00869853, 50 mg
SDP
Catalog No. 501011667
Encompass_Preferred
MF: C44H69NO12, MW: 804.02, Solubility: 26.6mg/mL in DMSO, 84.5 mg/mL in EtOH. Tacrolimus (FK506) is a potent and selective inhibitor of T-lymphocyte and the macrolide immunosuppressant.
Catalog No. 50-101-1667 Supplier Apexbio Technology LLC Supplier No. B214350
MF: C44H69NO12, MW: 804.02, Solubility: 26.6mg/mL in DMSO, 84.5 mg/mL in EtOH. Tacrolimus (FK506) is a potent and selective inhibitor of T-lymphocyte and the macrolide immunosuppressant.