Learn More
MOLECULAR DEPOT LLC Tacrolimus mAb Beads - 1 mL
Catalog No. 5000013513
Encompass_Preferred
Tacrolimus Antibody Conjugated to Magnetic Beads consists of magnetic particles (1 um in diameter 0 3perc solid) conjugated to a Tacrolimus mouse monoclonal antibody though a (PEG)n linker This product has been used as tools for the research and development of CLIA methods for the detection of Tacrolimus This product is for in vitro research use only
Catalog No. 50-000-13513 Supplier MOLECULAR DEPOT LLC Supplier No. B2010729
Tacrolimus Antibody Conjugated to Magnetic Beads consists of magnetic particles (1 um in diameter 0 3perc solid) conjugated to a Tacrolimus mouse monoclonal antibody though a (PEG)n linker This product has been used as tools for the research and development of CLIA methods for the detection of Tacrolimus This product is for in vitro research use only