Learn More
Selleck Chemical LLC Tacrolimus monohydrate
SDP
Catalog No. 5000003256
Encompass_Preferred
Tacrolimus monohydrate (Tacrolimus FK506) is a macrolide antibiotic that primarily acts as an inhibitor of calcineurin phosphatase by binding to FK506 binding protein (FKBP) This inhibition blocks calcium-dependent processes including interleukin-2 gene transcription nitric oxide synthase activation cell degranulation and apoptosis and also has immunosuppressive neuroregenerative and neuroprotective properties
Catalog No. 50-000-03256 Supplier Selleck Chemical LLC Supplier No. S5788100MG
Tacrolimus monohydrate (Tacrolimus FK506) is a macrolide antibiotic that primarily acts as an inhibitor of calcineurin phosphatase by binding to FK506 binding protein (FKBP) This inhibition blocks calcium-dependent processes including interleukin-2 gene transcription nitric oxide synthase activation cell degranulation and apoptosis and also has immunosuppressive neuroregenerative and neuroprotective properties