Learn More
Apexbio Technology LLC Tacrolimus monohydrate 109581-93-3 100mg
SDP
Catalog No. 5000062698
Encompass_Preferred
Tacrolimus monohydrate also identified as FK506 monohydrate is a macrolide immunosuppressive compound functioning via inhibition of calcineurin-mediated signaling pathways Specifically tacrolimus forms a complex with FK506-binding protein (FKBP12) subsequently blocking calcineurin phosphatase activity thus suppressing T-cell activation and diminishing interleukin-2 (IL-2) transcription levels In cellular assays tacrolimus demonstrates inhibition of calcineurin with IC50 values typically ranging between 0 1-1 0 nM Due to its targeted modulation of immune signaling this compound remains widely utilized in biomedical research particularly for investigations related to immune response mechanisms autoimmune disorder models transplantation biology and signal transduction pathways relevant to lymphocyte activation and cytokine expression
Catalog No. 50-000-62698 Supplier Apexbio Technology LLC Supplier No. B3579100
Tacrolimus monohydrate also identified as FK506 monohydrate is a macrolide immunosuppressive compound functioning via inhibition of calcineurin-mediated signaling pathways Specifically tacrolimus forms a complex with FK506-binding protein (FKBP12) subsequently blocking calcineurin phosphatase activity thus suppressing T-cell activation and diminishing interleukin-2 (IL-2) transcription levels In cellular assays tacrolimus demonstrates inhibition of calcineurin with IC50 values typically ranging between 0 1-1 0 nM Due to its targeted modulation of immune signaling this compound remains widely utilized in biomedical research particularly for investigations related to immune response mechanisms autoimmune disorder models transplantation biology and signal transduction pathways relevant to lymphocyte activation and cytokine expression