Learn More
TARGETMOL CHEMICALS INC Tacrolimus monohydrate 100MG
SDP
Catalog No. 502875970
Encompass_Preferred
Also available in 1 mL, 5 mg, 10 mg, 50 mg, 500 mg and bulk. Please contact Fisher for quotes. Tacrolimus monohydrate (LCP-Tacro) is a macrolide from the culture broth of a strain of Streptomyces tsukubaensis, binds to FK506 binding protein (FKBP) to form a complex. Tacrolimus inhibits calcineurin phosphatase, which inhibits T-lymphocyte signal transduction and IL-2 transcription. Immunosuppressive properties. It prevents the activation of T-lymphocytes in response to antigenic or mitogenic stimulation in vitro and has strong immunosuppressive activity in vivo. Purity 99.2%
Catalog No. 50-287-5970 Supplier TARGETMOL CHEMICALS INC Supplier No. T20879100MG
Also available in 1 mL, 5 mg, 10 mg, 50 mg, 500 mg and bulk. Please contact Fisher for quotes. Tacrolimus monohydrate (LCP-Tacro) is a macrolide from the culture broth of a strain of Streptomyces tsukubaensis, binds to FK506 binding protein (FKBP) to form a complex. Tacrolimus inhibits calcineurin phosphatase, which inhibits T-lymphocyte signal transduction and IL-2 transcription. Immunosuppressive properties. It prevents the activation of T-lymphocytes in response to antigenic or mitogenic stimulation in vitro and has strong immunosuppressive activity in vivo. Purity 99.2%