Learn More
Medchemexpress LLC Tacrolimus monohydrate | 109581-93-3 | 99.9% | 822.03 | 10 MM 1 ML
SDP
Catalog No. 5000352277
Encompass_Preferred
Tacrolimus monohydrate is a macrocyclic lactone that binds to FKBP to form a complex, inhibiting calcineurin phosphatase. This action suppresses T-lymphocyte signal transduction and IL-2 transcription, resulting in immunosuppressive properties. It is intended for research use only.
  • Inhibits T-lymphocyte signal transduction
  • Inhibits IL-2 transcription
  • Immunosuppressive properties
  • Potentiates glucocorticoid and progesterone actions
  • May enhance TGFβ-1 gene expression
  • Inhibits T cell proliferation
Catalog No. 50-003-52277 Supplier Medchemexpress LLC Supplier No. HY13756A10MM/1ML
Tacrolimus monohydrate is a macrocyclic lactone that binds to FKBP to form a complex, inhibiting calcineurin phosphatase. This action suppresses T-lymphocyte signal transduction and IL-2 transcription, resulting in immunosuppressive properties. It is intended for research use only.
  • Inhibits T-lymphocyte signal transduction
  • Inhibits IL-2 transcription
  • Immunosuppressive properties
  • Potentiates glucocorticoid and progesterone actions
  • May enhance TGFβ-1 gene expression
  • Inhibits T cell proliferation