Learn More
Medchemexpress LLC Tacrolimus monohydrate | 109581-93-3 | 99.9% | 822.03 | 200 MG
SDP
Catalog No. 5000352012
Encompass_Preferred
Tacrolimus monohydrate is a macrocyclic lactone that binds to FK506 binding protein (FKBP) to form a complex. This complex inhibits calcineurin phosphatase, which in turn inhibits T-lymphocyte signal transduction and IL-2 transcription. It exhibits immunosuppressive properties and is for research use only.
  • Binds to FK506 binding protein (FKBP) to form a complex
  • Inhibits calcineurin phosphatase
  • Inhibits T-lymphocyte signal transduction and IL-2 transcription
  • Exhibits immunosuppressive properties
  • Inhibits calcium-dependent events such as IL-2 gene transcription, NO synthase activation, cell degranulation, and apoptosis
  • Potentiates glucocorticoid and progesterone actions
  • May enhance TGFβ-1 gene expression
  • Inhibits T cell proliferation
Catalog No. 50-003-52012 Supplier Medchemexpress LLC Supplier No. HY13756A200MG
Tacrolimus monohydrate is a macrocyclic lactone that binds to FK506 binding protein (FKBP) to form a complex. This complex inhibits calcineurin phosphatase, which in turn inhibits T-lymphocyte signal transduction and IL-2 transcription. It exhibits immunosuppressive properties and is for research use only.
  • Binds to FK506 binding protein (FKBP) to form a complex
  • Inhibits calcineurin phosphatase
  • Inhibits T-lymphocyte signal transduction and IL-2 transcription
  • Exhibits immunosuppressive properties
  • Inhibits calcium-dependent events such as IL-2 gene transcription, NO synthase activation, cell degranulation, and apoptosis
  • Potentiates glucocorticoid and progesterone actions
  • May enhance TGFβ-1 gene expression
  • Inhibits T cell proliferation