Learn More
Medchemexpress LLC Tacrolimus monohydrate | 109581-93-3 | 99.93% | 822.03 | 1 G
SDP
Catalog No. 5000351845
Encompass_Preferred
Tacrolimus monohydrate, a macrocyclic lactone, forms a complex by binding to FK506 binding protein (FKBP). This complex then inhibits calcineurin phosphatase, which in turn inhibits T-lymphocyte signal transduction and IL-2 transcription, leading to immunosuppressive properties. It also inhibits calcium-dependent events like IL-2 gene transcription, NO synthase activation, cell degranulation, and apoptosis. Tacrolimus can potentiate the actions of glucocorticoids and progesterone by binding to FKBPs within the hormone receptor complex, preventing their degradation. The agent may also enhance the expression of the TGFβ-1 gene similarly to CsA, and it inhibits T cell proliferation in response to T cell receptor ligation.
  • Enhances expression of the TGFβ-1 gene
  • Inhibits T cell proliferation in response to T cell receptor ligation
  • Immunosuppressive properties
  • Inhibits calcineurin phosphatase
Catalog No. 50-003-51845 Supplier Medchemexpress LLC Supplier No. HY13756A1G
Tacrolimus monohydrate, a macrocyclic lactone, forms a complex by binding to FK506 binding protein (FKBP). This complex then inhibits calcineurin phosphatase, which in turn inhibits T-lymphocyte signal transduction and IL-2 transcription, leading to immunosuppressive properties. It also inhibits calcium-dependent events like IL-2 gene transcription, NO synthase activation, cell degranulation, and apoptosis. Tacrolimus can potentiate the actions of glucocorticoids and progesterone by binding to FKBPs within the hormone receptor complex, preventing their degradation. The agent may also enhance the expression of the TGFβ-1 gene similarly to CsA, and it inhibits T cell proliferation in response to T cell receptor ligation.
  • Enhances expression of the TGFβ-1 gene
  • Inhibits T cell proliferation in response to T cell receptor ligation
  • Immunosuppressive properties
  • Inhibits calcineurin phosphatase