Learn More
Medchemexpress LLC HY-13756A 50mg , Tacrolimus (monohydrate) Fujimycin monohydrate;FR900506 monohydrate;FK506 monohydrate CAS:109581-93-3 Purity:98%
SDP
Supplier:  Medchemexpress LLC HY13756A50MG
Encompass_Preferred
Medchemexpress, HY-13756A 50mg Tacrolimus (monohydrate) Fujimycin monohydrate;FR900506 monohydrate;FK506 monohydrate CAS:109581-93-3 Formula:C44H71NO13 PP2B (calcineurin phosphatase) Autophagy inducer Purity:98% Tacrolimus monohydrate binds to FK506 binding protein ( FKBP ). This complex inhibits calcineurin phosphatase (PP2B). Tacrolimus monohydrate is a mTOR-independent autophagy inducer. Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-149-3955