Learn More
Medchemexpress LLC Tacrolimus (Standard) | 104987-11-3 | 99.6% | 804.02 | 100 MG
SDP
Catalog No. 5000351846
Encompass_Preferred
Tacrolimus (Standard) is an analytical standard intended for research and analytical applications. This macrocyclic lactone binds to FK506 binding protein (FKBP) to form a complex. It inhibits calcineurin phosphatase, which in turn inhibits T-lymphocyte signal transduction and IL-2 transcription, possessing immunosuppressive properties.
  • Analytical standard for research and analytical applications
  • Used in qualitative research experiments
  • Used in quantitative research experiments
  • Used in methodological research experiments
  • Suitable for HPLC, GC, and MS applications
  • Inhibits calcineurin phosphatase
  • Inhibits T-lymphocyte signal transduction and IL-2 transcription
  • Possesses immunosuppressive properties
Catalog No. 50-003-51846 Supplier Medchemexpress LLC Supplier No. HY13756R100MG
Tacrolimus (Standard) is an analytical standard intended for research and analytical applications. This macrocyclic lactone binds to FK506 binding protein (FKBP) to form a complex. It inhibits calcineurin phosphatase, which in turn inhibits T-lymphocyte signal transduction and IL-2 transcription, possessing immunosuppressive properties.
  • Analytical standard for research and analytical applications
  • Used in qualitative research experiments
  • Used in quantitative research experiments
  • Used in methodological research experiments
  • Suitable for HPLC, GC, and MS applications
  • Inhibits calcineurin phosphatase
  • Inhibits T-lymphocyte signal transduction and IL-2 transcription
  • Possesses immunosuppressive properties