Learn More
Medchemexpress LLC Tacrolimus (standard) | 104987-11-3 | 99.6% | 804.02 | 25 MG
SDP
Catalog No. 5000352128
Encompass_Preferred
Tacrolimus (Standard) is the analytical standard of Tacrolimus, intended for research and analytical applications. This macrocyclic lactone binds to FK506 binding protein (FKBP) to form a complex, inhibiting calcineurin phosphatase, T-lymphocyte signal transduction, and IL-2 transcription, thereby exhibiting immunosuppressive properties.
  • Analytical standard for research and analytical applications.
  • Binds to FKBP and inhibits calcineurin phosphatase.
  • Inhibits T-lymphocyte signal transduction and IL-2 transcription.
  • Exhibits immunosuppressive properties.
  • Used as a reference standard in assays.
  • Suitable for qualitative, quantitative, and methodological research experiments in HPLC, GC, and MS.
Catalog No. 50-003-52128 Supplier Medchemexpress LLC Supplier No. 25MGHY13756R
Tacrolimus (Standard) is the analytical standard of Tacrolimus, intended for research and analytical applications. This macrocyclic lactone binds to FK506 binding protein (FKBP) to form a complex, inhibiting calcineurin phosphatase, T-lymphocyte signal transduction, and IL-2 transcription, thereby exhibiting immunosuppressive properties.
  • Analytical standard for research and analytical applications.
  • Binds to FKBP and inhibits calcineurin phosphatase.
  • Inhibits T-lymphocyte signal transduction and IL-2 transcription.
  • Exhibits immunosuppressive properties.
  • Used as a reference standard in assays.
  • Suitable for qualitative, quantitative, and methodological research experiments in HPLC, GC, and MS.