Learn More
IBA LifeSciences TACS Column Adapter; 1 adapter (for 0.3 ml Strep-Tactin TACS Agarose columns)
Catalog No. 502100165
Encompass
The adapters allow fitting of Strep-Tactin TACS Agarose columns onto standard tubes. Column adapters are available in two sizes: For 3 ml columns (containing 0.3 ml Strep-Tactin TACS Agarose) to fit them onto 15 ml tubes and for 10 ml columns (containing 1 ml Strep-Tactin TACS Agarose) to fit them onto 50 ml tubes.
Catalog No. 50-210-0165 Supplier IBA LifeSciences Supplier No. 63333001
The adapters allow fitting of Strep-Tactin TACS Agarose columns onto standard tubes. Column adapters are available in two sizes: For 3 ml columns (containing 0.3 ml Strep-Tactin TACS Agarose) to fit them onto 15 ml tubes and for 10 ml columns (containing 1 ml Strep-Tactin TACS Agarose) to fit them onto 50 ml tubes.