Learn More
IBA LifeSciences TACS Column Adapter
Catalog No. NC3566475
Encompass
The adapters allow fitting of Strep-Tactin TACS Agarose columns onto standard tubes. Column adapters are available in two sizes: for 0.3 ml Strep-Tactin TACS Agarose columns to fit them onto 15 ml tubes and for 1 ml Strep-Tactin TACS Agarose columns to fit them onto 50 ml tubes.
Catalog No. NC3566475 Supplier IBA LifeSciences Supplier No. 66331001
Add to Cart
Add to Cart
The adapters allow fitting of Strep-Tactin TACS Agarose columns onto standard tubes. Column adapters are available in two sizes: for 0.3 ml Strep-Tactin TACS Agarose columns to fit them onto 15 ml tubes and for 1 ml Strep-Tactin TACS Agarose columns to fit them onto 50 ml tubes.