Learn More
Aviva Systems Biology Corp TADA2L Antibody - middle region (ARP38045_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP38045_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against TADA2L. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human TADA2L. Alternate names: ADA2, FLJ12705, KL04P, hADA2, ADA2A, TADA2LApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Zebrafish
Catalog No. 50-014-56500