Learn More
Aviva Systems Biology Corp TADA3L Antibody - middle region (ARP31479_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP31479_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against TADA3L. It was validated on Western Blot and immunohistochemistry. The immunogen is a synthetic peptide directed towards the middle region of human TADA3L. Alternate names: ADA3, FLJ20221, FLJ21329, hADA3, NGG1, TADA3LApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Zebrafish
Catalog No. 50-014-54242