Learn More
Aviva Systems Biology Corp TADA3L Antibody - C-terminal region (ARP38786_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP38786_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against TADA3L. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the C terminal region of human TADA3L. Alternate names: ADA3, NGG1, hADA3, TADA3LApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Zebrafish
Catalog No. 50-014-56797