Learn More
AdipoGen Tadalafil (500 mg)
SDP
Supplier:  AdipoGen CDXT0455M500
Encompass_Preferred
Chemical. CAS: 171596-29-5. Formula: C22H19N3O4. MW: 389.40. Tadalafil is a potent long-acting and selective inhibitor of the cGMP specific phosphodiesterase 5 (PDE5; IC50 = 1.2 nM). It is selective for PDE5 over PDE1-4 and 7-10 (IC50s = 9.2-280 µM), however, it does also inhibit PDE11 (IC50 = 11 nM). In vivo, tadalafil (10 mg/kg) decreases production of the proinflammatory cytokines TNF-alpha, IL-1beta, and IL-6 and improves renal function in a rat model of ischemia/reperfusion injury. Formulations containing tadalafil have been used to treat erectile dysfunction, pulmonary arterial hypertension, benign prostatic hyperplasia (BPH) and lower urinary tract dysfunction. Also delays tumor growth in a mouse model via inhibition of MDSC-mediated immunosuppression.
Catalog No. 50-205-4671