Learn More
Apexbio Technology LLC Tadalafil, 50mg. Cas: 171596-29-5 MFCD: MFCD07771966. PDE5 inhibitor
SDP
Supplier:  Apexbio Technology LLC A432750
Encompass_Preferred
Tadalafil, also known as IC351 or Cialis, is a potent, reversible and competitive small-molecule inhibitor of phosphodiesterase 5 (PDE5), a predomiant PDE in the pulmonary vasculature that hydrolyzes cyclic guanosine monophosphate (cGMP). Other sizes are available. Please inqury us for quote.
Catalog No. 50-190-4924