Learn More
Cayman Chemical Tadalafil-d3, Purity: ≥99% deuterated forms (d1-d3), CAS: 960226-55-5, 1 mg
SDP
Catalog No. 502934510
Encompass_Preferred
An internal standard for the quantification of tadalafil by GC- or LC-MS
Catalog No. 50-293-4510 Supplier Cayman Chemical Supplier No. 250301 MG
An internal standard for the quantification of tadalafil by GC- or LC-MS