Learn More
Selleck Chemical LLC Tadalafil 50mg 171596-29-5 IC351
SDP
Catalog No. 506354
Encompass_Preferred
Tadalafil (IC351) is a PDE-5 inhibitor with IC50 of 1.8 nM in a cell-free assay. Tadalafil is at least 9000 times more selective for PDE5 than most of the other families of PDEs, with the exception of PDE11. It can partial inhibits PDE11 *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. 50-635-4 Supplier Selleck Chemical LLC Supplier No. S151250MG
Tadalafil (IC351) is a PDE-5 inhibitor with IC50 of 1.8 nM in a cell-free assay. Tadalafil is at least 9000 times more selective for PDE5 than most of the other families of PDEs, with the exception of PDE11. It can partial inhibits PDE11 *For Research & Development use only.  Product is not intended for drug, household, or human consumption.