Learn More
Strem, An Ascensus Company 100mg. (3aS,8aS)-Tetrahydro-6-hydroxy-2,2-dimethyl-4,4,8,8-tetrakis(3,5-diisopropyl-4-methoxyphenyl)-6-oxide-[1,3]dioxolo[4,5-e][1,3,2]dioxaphosphepine, 98%.
SDP
Supplier:  Strem, An Ascensus Company 158613100MG
Encompass
100mg. (3aS,8aS)-Tetrahydro-6-hydroxy-2,2-dimethyl-4,4,8,8-tetrakis(3,5-diisopropyl-4-methoxyphenyl)-6-oxide-[1,3]dioxolo[4,5-e][1,3,2]dioxaphosphepine, 98%. . Molecular Weight: 985.3. Molecular Formula: C59H85O10P. Color/form: off white to light yellow pwdr. Strem# 15-8613. http://www.strem.com/catalog/v/15-8613/
Catalog No. 50-206-5512