Learn More
Strem, An Ascensus Company 25mg. (3aR,8aR)-Tetrahydro-6-hydroxy-2,2-dimethyl-4,4,8,8-tetrakis(3,5-di-tert-butylphenyl)-6-oxide-[1,3]dioxolo[4,5-e][1,3,2]dioxaphosphepine, 98%, (99% ee).
SDP
Supplier:  Strem, An Ascensus Company 15861725MG
Encompass
25mg. (3aR,8aR)-Tetrahydro-6-hydroxy-2,2-dimethyl-4,4,8,8-tetrakis(3,5-di-tert-butylphenyl)-6-oxide-[1,3]dioxolo[4,5-e][1,3,2]dioxaphosphepine, 98%, (99% ee). . Molecular Weight: 977.4. Molecular Formula: C63H93O6P. Color/form: off white to light yellow pwdr. Strem# 15-8617. http://www.strem.com/catalog/v/15-8617/
Catalog No. 50-206-5513