Learn More
Medchemexpress LLC TAE-1 10mg | 1414469-59-2 | 1128.57 | C39H51I3N6O9 | 10 MG
SDP
Catalog No. 5000307945
Encompass_Preferred
TAE-1 is a small-molecule research compound that potently inhibits acetylcholinesterase (AChE) and butyrylcholinesterase (BuChE) and reduces amyloid-β (Aβ) fibril formation and aggregation, making it useful for in vitro and preclinical studies related to Alzheimer's disease.
  • High potency against AChE and BuChE.
  • Inhibits amyloid-β fibril formation and aggregation.
  • Supplied as a solid with reported purity of 96.17%.
  • Soluble in DMSO at 125 mg/mL (110.76 mM) with ultrasonic warming and heating to 60°C recommended.
  • Stable under recommended storage conditions for research use.
Catalog No. 50-003-07945 Supplier Medchemexpress LLC Supplier No. HY11565010MG
TAE-1 is a small-molecule research compound that potently inhibits acetylcholinesterase (AChE) and butyrylcholinesterase (BuChE) and reduces amyloid-β (Aβ) fibril formation and aggregation, making it useful for in vitro and preclinical studies related to Alzheimer's disease.
  • High potency against AChE and BuChE.
  • Inhibits amyloid-β fibril formation and aggregation.
  • Supplied as a solid with reported purity of 96.17%.
  • Soluble in DMSO at 125 mg/mL (110.76 mM) with ultrasonic warming and heating to 60°C recommended.
  • Stable under recommended storage conditions for research use.