Learn More
Medchemexpress LLC TAE-1 25mg | 1414469-59-2 | 25 MG
SDP
Catalog No. 5000226268
Encompass_Preferred
TAE-1 is a small-molecule inhibitor of acetylcholinesterase (AChE) and butyrylcholinesterase (BuChE) that also inhibits amyloid-β fibril formation and aggregation. It is used in biochemical and cellular research into Alzheimer's disease, cholinesterase function, and amyloid-β aggregation.
  • Potent inhibitor of AChE and BuChE.
  • Inhibits amyloid-β fibril formation and aggregation.
  • Molecular formula C39H51I3N6O9 and molecular weight 1128.57.
  • Typical supplied purity about 96.17%.
  • Available in small laboratory pack sizes (for example, 25 mg) for research use.
Catalog No. 50-002-26268 Supplier Medchemexpress LLC Supplier No. HY11565025MG
Add to Cart
Add to Cart
TAE-1 is a small-molecule inhibitor of acetylcholinesterase (AChE) and butyrylcholinesterase (BuChE) that also inhibits amyloid-β fibril formation and aggregation. It is used in biochemical and cellular research into Alzheimer's disease, cholinesterase function, and amyloid-β aggregation.
  • Potent inhibitor of AChE and BuChE.
  • Inhibits amyloid-β fibril formation and aggregation.
  • Molecular formula C39H51I3N6O9 and molecular weight 1128.57.
  • Typical supplied purity about 96.17%.
  • Available in small laboratory pack sizes (for example, 25 mg) for research use.