Learn More
Research Products International Corp TAE, 10 L

Supplier: Research Products International Corp T6001010000.0
TAE 10X Solution is a concentrated Tris-acetate-EDTA buffer used in molecular biology for DNA and RNA electrophoresis. When diluted to 1X, it maintains pH (~8.3) and low ionic strength. It supports clear nucleic acid separation, extended run times, and protects samples from degradation by chelating divalent metal ions like Mg²⁺.
Substance Name: [TRIS Acetate EDTA 10X Solution]
10X solution contains:
TRIS-Base: 0.4 M
Acetic Acid: 0.2 M
EDTA: 10 mM
pH (10X Solution (@25°C): 8.2-8.4
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.