Learn More
Kd Medical Inc TAE 10X 1L BOTTLE
SDP
Catalog No. 501031410
Encompass_Preferred
TAE Buffer (Tris Actetate EDTA) is the most commonly used buffer for agarose DNA electrophoresis. 10X concentrated ready to be diluted to a 1X working solution. One 1 liter bottle.
Catalog No. 50-103-1410 Supplier Kd Medical Inc Supplier No. RGF3310
TAE Buffer (Tris Actetate EDTA) is the most commonly used buffer for agarose DNA electrophoresis. 10X concentrated ready to be diluted to a 1X working solution. One 1 liter bottle.