Learn More
Kd Medical Inc TAE 50X, 1L BOTTLE
SDP
Catalog No. 501031411
Encompass
TAE Buffer (Tris Actetate EDTA) is the most commonly used buffer for agarose DNA electrophoresis. 50X concentrated ready to be diluted to a 1X working solution. One 1 liter bottle.
Catalog No. 50-103-1411 Supplier Kd Medical Inc Supplier No. RGF3320
TAE Buffer (Tris Actetate EDTA) is the most commonly used buffer for agarose DNA electrophoresis. 50X concentrated ready to be diluted to a 1X working solution. One 1 liter bottle.