Learn More
Research Products International Corp TAE 50X Buffer [TRIS Acetate EDTA 50X Solution], 1 Liter
SDP
Catalog No. 50489104
Encompass_Preferred
TAE 50X Buffer is a concentrated solution of Tris, acetic acid, and EDTA used in nucleic acid electrophoresis. When diluted, it maintains pH (~8.3) and low ionic strength, supporting clear DNA/RNA separation. EDTA chelates metal ions, helping prevent nucleic acid degradation during electrophoretic analysis in molecular biology workflows.
Substance Name: TRIS Acetate EDTA 50X Solution
Solution contains:
Tris (hydroxymethyl) aminomethane: 2 M
Acetic Acid: 1 M
EDTA, Disodium Salt Dihydrate: 50 mM
Catalog No. 50-489-104 Supplier Research Products International Corp Supplier No. T600151000.0
Add to Cart
Add to Cart
TAE 50X Buffer is a concentrated solution of Tris, acetic acid, and EDTA used in nucleic acid electrophoresis. When diluted, it maintains pH (~8.3) and low ionic strength, supporting clear DNA/RNA separation. EDTA chelates metal ions, helping prevent nucleic acid degradation during electrophoretic analysis in molecular biology workflows.
Substance Name: TRIS Acetate EDTA 50X Solution
Solution contains:
Tris (hydroxymethyl) aminomethane: 2 M
Acetic Acid: 1 M
EDTA, Disodium Salt Dihydrate: 50 mM