Learn More
Growcells TAE BUFFER 10X 2L EA
Catalog No. 501034826
Encompass_Preferred
Sterile liquid electrophoresis buffer concentrate prepared with 18.2 megOhm water, 0.2 micron sterile filtered and steam sterilized in final packaging. Recipe/L: 48.46g TRIS Base, 11.4ml Acetic Acid, 3.72g EDTA. pH 8.3
Catalog No. 50-103-4826 Supplier Growcells Supplier No. MRGF4201
Sterile liquid electrophoresis buffer concentrate prepared with 18.2 megOhm water, 0.2 micron sterile filtered and steam sterilized in final packaging. Recipe/L: 48.46g TRIS Base, 11.4ml Acetic Acid, 3.72g EDTA. pH 8.3