Learn More
Crescent Chemical Co Inc TAE BUFFER (10X) MOL BIO GRADE
SDP
Catalog No. 50248606
Encompass_Preferred
TAE buffer (10x) molecular biology grade 10 x concentrated aqueous solution.TAE Buffer is used for the electrophoresis of nucleic acids. TAE has a lower buffer capacity than TBE, however linear ds DNA tends to run faster in TAE than in TBE. Size - 1L Stor
Catalog No. 50-248-606 Supplier Crescent Chemical Co Inc Supplier No. SE42553.01
TAE buffer (10x) molecular biology grade 10 x concentrated aqueous solution.TAE Buffer is used for the electrophoresis of nucleic acids. TAE has a lower buffer capacity than TBE, however linear ds DNA tends to run faster in TAE than in TBE. Size - 1L Stor